Learn More
Waters Corp Waters XBridge Preparative Column, BEH Amide, 5 μm, 30 x 50 mm, OBD
Supplier:  Waters Corp 186006607
Encompass_Preferred
OBD Preparative Columns bridge the performance gap in Prep LC, offering uniform, optimally packed bed density throughout the column. They provide direct scalability from analytical to preparative chromatography, excellent column lifetime, and consistent column-to-column performance for good resolution, peak shape, and mass loading. This technology delivers on-target results for successful preparative LC purification.
  • OBD column type ensures uniform, optimally packed bed density.
  • BEH Amide chemistry for specific separation needs.
  • Particle size of 5 μm.
  • Dimensions of 30 x 50 mm.
  • Provides direct scalability from analytical to preparative chromatography.
  • Offers excellent column lifetime.
  • Delivers consistent column-to-column performance.
  • Achieves good resolution, peak shape, and mass loading.
Catalog No. 50-466-360