Learn More
Waters Corp Waters OBD Prep Column, BEH C18, 10 μm, 30 mm X 100 mm, 130 Å
Supplier:  Waters Corp 186003930
Encompass_Preferred
This OBD Prep Column uses BEH C18 sorbent for HPLC method development, functional across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand offers a versatile column for HPLC, with superior low pH stability and ultra-low column bleed.
  • Uniform, optimally packed column beds.
  • Direct scalability from analytical to preparative liquid chromatography.
  • Extended column lifetime.
  • Consistent column-to-column performance.
  • Good resolution, peak shape, and mass loading.
  • OBD Technology for successful Prep LC purification.
  • Universal column choice for various compounds.
  • Exceptional peak shape for basic analytes at elevated pH.
  • Stable across a wide pH range and high temperatures.
  • Full Coverage C18 provides balanced non-polar retention.
  • High loading capacity.
  • BEH particle option for maximum pH stability.
Catalog No. 50-782-067
Add to Cart
Request a Quote