Learn More
Waters Corp Waters XBridge Preparative Column, BEH C8, 30 x 75 mm, 5 μm
Supplier:  Waters Corp 186003269
Encompass_Preferred
The XBridge Preparative Column, BEH C8, is a versatile column designed for a wide range of chromatographic applications. It offers stable performance across varying pH and temperatures, making it suitable for separating diverse compounds, including strongly hydrophobic ones.
  • Good universal column choice for a variety of compounds.
  • Exceptional peak shape for basic analytes at elevated pH.
  • Stable across a wide pH range.
  • Stable at high temperatures.
  • Shorter alkyl chain providing less retentivity than a typical C18.
  • Suitable for the retention of strongly hydrophobic compounds.
Catalog No. 50-781-641
Add to Cart
Request a Quote