Learn More
Waters Corp Waters OBD Prep Column, BEH Phenyl, 5 μm, 30 mm X 150 mm
Supplier:  Waters Corp 186003276
Encompass_Preferred
This preparative column offers complementary selectivity compared to C18 stationary phases, particularly for polyaromatic compounds. It features the most chemically stable commercially available Phenyl sorbent on the market, providing ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Most chemically stable commercially available Phenyl sorbent.
  • Provides ultra-low column bleed.
  • Offers long column lifetimes.
  • Delivers excellent peak shape.
Catalog No. 50-781-647
Add to Cart
Request a Quote