Learn More
Waters Corp Column, 300 Å, 2.5 μm, 2.1 mm x 100 mm
Supplier:  Waters Corp 186009128
Encompass_Preferred
These columns are optimized and quality control tested for protein separations. They feature a 300Å pore size trifunctionally bonded BEH particle, which offers a wide usable pH range, superior low pH stability, and ultra-low column bleed for various protein-based applications.
  • Optimized and quality control tested for protein separations using a protein mix standard.
  • Trifunctionally bonded BEH particle with 300Å pore size.
  • Offers the widest usable pH range.
  • Provides superior low pH stability.
  • Ensures ultra-low column bleed.
  • Suggested for low-level intact protein and subunit analysis.
Catalog No. 50-239-2638