Learn More
Waters Corp Preparative Column, C4, 10 μm, 300 Å, 19 mm x 150 mm
Supplier:  Waters Corp 186007333
Encompass_Preferred
These preparative columns are quality control tested and optimized to separate proteins based on size, hydrophobicity, and isoelectric point. The C4 ligand offers less retention compared to traditional C18 phases, which helps to minimize protein carryover, increase protein recovery, and improve peak capacity.
  • QC tested for protein separation by size, hydrophobicity, and isoelectric point.
  • C4 ligand reduces protein carryover, increases recovery, and improves peak capacity.
  • Employs BEH technology and 300 Å packing for excellent performance with biological macromolecules.
  • Offers stability across a wide pH range and high temperatures.
  • Manufactured in a cGMP, ISO 9002 accredited facility; each batch tested with a certificate of analysis.
Catalog No. 50-239-2182