Learn More
Waters Corp Preparative Column, C4, 300 Å Pore, 10 μm Particle, 30 mm × 250 mm
Supplier:  Waters Corp 186007340
Encompass_Preferred
XBridge Protein BEH C4 OBD Columns are quality control tested and optimized for the separation of proteins based on size, hydrophobicity, and isoelectric point. Compared to traditional C18 phases, the C4 ligand offers less retention, which helps to minimize protein carryover, increase protein recovery, and improve peak capacity.
  • Optimized for protein separation based on size, hydrophobicity, and isoelectric point.
  • C4 ligand provides less retention compared to traditional C18 phases.
  • Minimizes protein carryover.
  • Increases protein recovery.
  • Improves peak capacity.
Catalog No. 50-239-2728
Add to Cart
Request a Quote