Learn More
Waters Corp Waters XBridge Protein BEH SEC Column, 450Å, 2.5 μm, 4.6 mm X 300 mm
Supplier:  Waters Corp 176004339
Encompass_Preferred
The XBridge Protein BEH SEC, 450Å Columns utilize 2.5 μm particles based on Ethylene Bridged Hybrid (BEH)-based particle technology and a diol-bonded surface coating. These columns are designed to separate proteins ranging from 100K to 1,500K Daltons.
  • Unique mobile-phase pH stability extends column lifetime.
  • Boosts productivity and supports quality control or LC-MS assay development.
  • Ensures excellent reliability and assay ruggedness.
  • Offers unmatched peak particle shape, capacity, and efficiency.
  • BEH particle technology with dial-bonded surface coating is ideal for high-range protein separation.
  • Operates ideally within a 2–8 pH range.
  • Includes one vial of the BEH450 Protein Standard for performance benchmarking.
  • Manufactured under strict ISO9001 and cGMP conditions.
Catalog No. 50-203-7591