Learn More
Waters Corp SEC Column, 200Å, 2.5 μm, 7.8 mm x 150 mm
Supplier:  Waters Corp 176004326
Encompass_Preferred
The XBridge Protein BEH SEC Column, 200Å, 2.5 μm, 7.8 mm X 150 mm, is designed to separate proteins ranging from 10,000 to 450,000 Daltons. This column utilizes Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating.
  • Offers unique mobile-phase pH stability to extend column lifetime.
  • Suitable for quality control techniques or to develop cutting-edge LC-MS assays.
  • Provides outstanding column reliability, ensuring assay ruggedness.
  • Delivers unrivaled peak particle shape and capacity with exceptional efficiency.
  • Includes one vial of the BEH200 SEC Protein Standard Mix at no additional cost.
  • Manufactured in accordance with ISO9001 and cGMP guidelines.
Catalog No. 50-203-7581