Learn More
Waters Corp Waters XBridge BEH Shield RP18 Column, 130Å, 3.5 μm, 2.1 mm, 150 mm
Supplier:  Waters Corp 186003038
Encompass_Preferred
This XBridge BEH Shield RP18 column for HPLC method development enhances selectivity for phenolic compounds and improves separation of basic analytes. Its proprietary Shield Technology minimizes silanol interactions, leading to superior peak symmetry and efficiency. With a wide pH stability (2–11), it offers robust, long-lasting performance for analytical and preparative applications.
  • Enhanced selectivity, especially for phenolic compounds.
  • Improved peak symmetry and efficiency via Shield Technology.
  • Superior performance over conventional C18 columns.
  • Robustness and longevity with wide pH stability (2–11 pH).
  • Versatile for HPLC, UPLC transfer, and preparative scaling.
  • A powerful tool for HPLC method development.
Catalog No. 50-781-491