Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XBridge Shield RP18 Column, 5 μm, 3.0 x 250 mm

Supplier:  Waters Corp 186003161

Encompass_Preferred

This product is a Waters XBridge Shield RP18 Column designed for various chromatography applications.

  • Features a 5 μm particle size.
  • Has dimensions of 3.0 x 250 mm.
  • Suitable for analytical chromatography.

Catalog No. 50-781-593


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.