Learn More
Waters Corp Waters HPLC Column, 130Å, 5 μm, 2.1 mm, 150 mm
Supplier:  Waters Corp 186003003
Encompass_Preferred
These BEH Shield RP18 columns are powerful tools for HPLC method development, complementing BEH C18 and C8 phases. They offer alternate selectivity for phenolic compounds due to an embedded carbamate group in the bonded phase ligand.
  • Wide and usable pH range for various analytes.
  • MaxPeak High-Performance Surface technology prevents unwanted analyte-surface interactions.
  • Ethylene-Bridged Hybrid (BEH) base particle ensures increased stability.
  • BEH particle from two high-purity monomers for stable, pH-resistant environment essential for method development.
  • Facilitates easy method transfer from HPLC to UHPLC or UPLC systems.
  • RP18 chemistry functions similarly to C18 or C8 base chemistries.
Catalog No. 50-781-456