Learn More
Waters Corp XBridge BEH Shield RP18 Column, 130Å, 5 µm, 4.6 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186003009
Encompass_Preferred
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
Catalog No. 50-781-462