Learn More
Waters Corp WATERS CORPORATION Column, XBridge Shield RP18, 2.5μm, 3.0x20mm, IS Cl
Supplier:  Waters Corp 186003140
Encompass_Preferred
This XBridge Shield RP18 Intelligent Speed Column is designed for chromatographic separations. It features a 2.5μm particle size and dimensions of 3.0x20mm, making it suitable for applications requiring high efficiency and rapid analysis.
  • Features XBridge Shield RP18 chemistry
  • 2.5μm particle size for high efficiency
  • 3.0x20mm column dimensions
  • Intelligent Speed (IS) for fast separations
  • Consumable product for laboratory use
Catalog No. 50-781-574