Learn More
Waters Corp Waters XBridge Peptide BEH C18 OBD Prep Column, 300Å, 5 μm, 10 mm x 250 mm
Supplier:  Waters Corp 186008193
Encompass_Preferred
XBridge Peptide BEH C18 columns are optimized and QC tested for peptide separations. Designed with a wide-pore (300Å) trifunctionally bonded BEH particle, this column provides a wide usable pH range, superior low pH stability, and ultra-low column bleed for proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Features wide-pore (300Å) trifunctionally bonded BEH particles.
  • Offers wide usable pH range (1–12) and superior low pH stability.
  • Ensures ultra-low column bleed.
  • Suitable for proteomics, protein characterization, and peptide synthesis.
  • Provides excellent peak shape, high efficiency, and stability.
  • Rigorous QC testing with peptide mapping.
  • C18 ligand ensures a stable, well-characterized baseline.
  • Achieves batch-to-batch consistency.
Catalog No. 50-138-4680