Learn More
Medchemexpress LLC XCL1 Rat His 2ug

Supplier: Medchemexpress LLC HYP71947A2UG
Recombinant rat XCL1 (His-tag) supplied as a lyophilized powder for in vitro biological assays, including chemotaxis and receptor activation studies. The protein is expressed in E. coli and characterized for purity, endotoxin, and activity to support reproducible experimental results.
- Recombinant rat chemokine with N-terminal 6xHis tag.
- Expressed in Escherichia coli and provided as lyophilized powder.
- Purity ≥95% by reducing SDS-PAGE.
- Endotoxin <1 EU/μg (LAL).
- Approximate molecular weight ~13 kDa (SDS-PAGE).
- Biological activity validated; ED50 ~27 ng/mL (chemotaxis assay).
- Stable stored at -20°C; follow reconstitution and storage recommendations for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.