Learn More
MSC Xcluder Rodent Fill Fabric, Stainless Steel/Fiber Blend, 4 in x 10 ft
Supplier:  MSC 74317066
Encompass
Xcluder Rodent Fill Fabric is a durable, non-toxic barrier made from a stainless steel and fiber blend. It is designed to seal gaps and block pests, preventing rodents from entering structures by compressing tightly into openings. This eco-friendly solution can be used indoors and outdoors around pipes, vents, and foundations.
  • Made from a durable stainless steel and fiber blend.
  • Compresses tightly and expands to fill gaps, cracks, and holes.
  • Creates an impenetrable, non-chemical barrier against pests.
  • Prevents gnawing from rodents like mice, rats, and chipmunks.
  • Supports Integrated Pest Management (IPM) best practices.
  • Safe for use around pets, children, and food.
  • Resistant to rust, unlike steel wool.
  • Provides superior protection over spray foam.
  • Offers fast, proven, and permanent pest exclusion.
Catalog No. 50-283-7299