Learn More
New England Biolabs, Inc. XcmI – 1000 units
SDP
Supplier:  New England Biolabs, Inc. R0533S
Encompass
A restriction endonuclease that recognizes the sequence CCANNNN_N^NNNNTGG.
Catalog No. 50-812-198