Learn More
Selleck Chemical LLC XCT790 S0407-5MG
SDP
Supplier:  Selleck Chemical LLC S04075MG
Encompass_Preferred
XCT-790 (Compound 12) is a potent and selective inverse agonist for estrogen-related receptor (ERR ) with IC50 of 0 37 M XCT-790 (Compound 12) is inactive against ERR and the estrogen receptors ER and ER XCT-790 (Compound 12) significantly inhibits in vivo tumor growth and angiogenesis and induces apoptosis
Catalog No. 50-000-41477