Learn More
MARSHALL SCIENTIFIC Edwards XDS35iC Dry Scroll Pump
SDP
Supplier:  MARSHALL SCIENTIFIC EDA73006983
Encompass_Preferred
Scroll pumps are an excellent alternative to rotary vane pumps where oil free pumping is desirable. The XDS pumps in particular use an innovative bearing shield to isolate the vacuum environment from all forms of lubricant, not only making them totally dry, but also protecting the bearing from any process gases. Vapour handling is enabled by an adjustable gas ballast; this allows the range of pumps to satisfy many applications that were previously off limits to other scroll pumps. Ultimate vacuum of 0.008 mbar and peak pumping speed of 21.0 CFM. “C” denotes this pump as the corrosion resistant variant.
Catalog No. NC2888781
Only null left
Add to Cart
Request a Quote