Learn More
Medchemexpress LLC Xe-991 dihydrochloride | 122955-42-4 | 99.7% | 376.45 Da | C26H20N2O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY108577A5MG
Encompass_Preferred
XE991 dihydrochloride is a potent, selective blocker of KCNQ (Kv7) voltage-gated potassium channels used in electrophysiology and pharmacology research to inhibit neuronal M-current. Supplied as a solid research reagent, reported purity is approximately 99.7%; vendors may list either the free base or the dihydrochloride salt form.
Catalog No. 50-002-64899