Learn More
Medchemexpress LLC Xevinapant | 1071992-99-8 | C32H43N5O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY15454500MG
Encompass_Preferred
Xevinapant is a potent and orally bioavailable Smac mimetic that acts as an antagonist of IAPs, binding to XIAP, cIAP1, and cIAP2 proteins.
  • Potent Smac mimetic
  • Orally bioavailable compound
  • Antagonizes IAP proteins
  • Binds to XIAP, cIAP1, and cIAP2
  • Used as a laboratory chemical
  • Suitable for manufacturing substances
Catalog No. 50-003-76471