Learn More
Waters Corp Waters Corporation Standard Kit, For Xevo TQD, Performance Verification
Supplier: Waters Corp 700006006
The Xevo TQD Standard Kit is designed for verifying the performance of the Xevo TQD, an advanced quadrupole mass detector for LC-MS/MS applications. This kit, suitable for both qualitative and quantitative applications, supports various ionization options. It requires refrigerated storage at 4°C, has a 12-month shelf life, and is classified as hazardous material.
- Intended for TQD performance verification.
- Detection method: MS.
- Formulation: Liquid and solid.
- Storage condition: 4°C.
- System type: LCMS.
- Shelf life: 12 months.
- Shipping condition: Ambient.
- Reference type.
- Application: System performance.
- Units per package: 1.
- Contains hazardous material.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.