Learn More
Agilent Technologies Seahorse XFe24 FluxPak. For use with the Seahorse XFe24 Analyzer. FluxPak includes 18 XFe24 sensor cartridges, 20 XF24 cell culture microplates, and 1 bottle of Seahorse XF Calibrant Solution 500ml.
Supplier: Agilent Technologies 102340100
Seahorse XFe24 FluxPak. For use with the Seahorse XFe24 Analyzer. FluxPak includes 18 XFe24 sensor cartridges, 20 XF24 cell culture microplates, and 1 bottle of Seahorse XF Calibrant Solution 500ml.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.