Learn More
Sorbent Technologies Inc Nano-Silica XHL HPTLC Plates, glass backed, 200um, 10x10cm, 25/pk
SDP
Supplier:  Sorbent Technologies Inc 4214056
Encompass_Preferred
This high performance thin layer chromatography (HPTLC) plate comprises a "Nano-Silica XHL" layer. The Silica 60 has a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, and particle size of 2-10um. This analytical extra hard layer is bound to a glass support. It has a thickness of 200um and dimensions of 10x10cm. There are 25 plates per pack. The layer has an outstanding hardness and is abrasion resistant due to an optimized binder system. The plate can be easily labeled with a lead pencil for clear identification. There is an increased separation efficiency due to an optimized particle size distribution.
Catalog No. NC2735675