Learn More
Apexbio Technology LLC XL-888, 5mg. Cas: 1149705-71-4 MFCD: MFCD22124888. Hsp90 inhibitor
SDP
Supplier:  Apexbio Technology LLC A43885
Encompass_Preferred
XL-888 is a novel and orally-bioavailable inhibitor of heat shock protein 90 (HSP90) that selectively inhibits HSP90 and HSP90 with values of 50% inhibition concentration IC50 of 22 nM and 44 nM respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4938