Learn More
Refrigiwear Inc. Iron-Tuff Inspector
SDP
Supplier:  Refrigiwear Inc. 0341RNAVXLG
Encompass_Preferred
The longest days, the harshest conditions, the toughest jobs - none of it is a match for this coat. Extra long for more protection, with side seam slits to access your pant pockets, this coat offers functionality beyond -50°F protection. The abrasion-resistant, water-resistant, wind-tight 400-denier nylon outer shell of this freezer jacket is strong and durable.
Catalog No. NC3404848