Learn More
Apexbio Technology LLC XL019, 5mg. Cas: 945755-56-6 MFCD: MFCD24386874. JAK2 inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A39375
Encompass_Preferred
XL019 is a potent and selective inhibitor of JAK2 with IC50 value of 2.2 nM.JAK kinases were first identified in 1989 and characterized by the presence of two tyrosine kinase domains. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4849