Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[4-[2-(4-morpholin-4-ylanilino)pyrimidin-4-yl]phenyl]pyrrolidine-2-carboxamide | 945755-56-6 | MFCD24386874 | 99.7% | 444.53 | C25H28N6O2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY137755MG

Encompass_Preferred

XL019 is a potent, orally bioavailable, selective inhibitor of Janus kinase 2 (JAK2) with low-nanomolar enzymatic potency and demonstrated cellular activity against JAK2-driven signaling.

  • Potent JAK2 inhibition (IC50 ≈ 2.2 nM) with weaker activity against JAK1 and JAK3.
  • Inhibits STAT3 and STAT5 phosphorylation in cells expressing mutant or wild-type JAK2.
  • High chemical purity (≈99.7%) suitable for research applications.
  • Molecular weight 444.53 and formula C25H28N6O2.
  • Soluble in DMSO (≈25 mg/mL); use fresh DMSO and ultrasonic assistance if needed.
  • Solid, light yellow to green yellow appearance; supplied as a 5 mg quantity.
  • Recommended storage: powder -20°C for long-term; in solution -80°C.

Catalog No. 50-208-1508


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.