Learn More
Selleck Chemical LLC XL092-E0142-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC E014210MM/1ML
Encompass_Preferred
XL092 (JUN04542) is an ATP-competitive inhibitor of multiple RTKs including MET VEGFR2 AXL and MER with IC50 values of 15 nM 1 6 nM 3 4 nM and 7 2 nM in cell-based assays respectively
Catalog No. 50-313-6875