Learn More
Agilent Technologies XL1-RED COMPETENT CELLS
Supplier:  Agilent Technologies 200129
Encompass
XL1-Red Competent Cells. For random mutagenesis, 1 x 106 transformants/ug, includes XL1-Blue competent cells. simple, quickly and easily introduce random mutations into gene of interest
Catalog No. 50-125-009