Learn More
Selleck Chemical LLC XL147 analogue 10mg 956958-53-5 SAR245408
SDP
Supplier:  Selleck Chemical LLC S111810MG
Encompass_Preferred
XL147 analogue (SAR245408) is a selective and reversible class I PI3K inhibitor for PI3K?/?/? with IC50 of 39 nM/36 nM/23 nM in cell-free assays, less potent to PI3K?. XL147 analogue induces apoptosis. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-879-4