Learn More
Selleck Chemical LLC Voxtalisib (XL765) Analogue 5mg 1349796-36-6 SAR245409
SDP
Supplier:  Selleck Chemical LLC S15235MG
Encompass_Preferred
Voxtalisib (SAR245409, XL765) Analogue is a dual inhibitor of mTOR/PI3K, mostly for p110? with IC50 of 9 nM; also inhibits DNA-PK and mTOR. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-820-3