Learn More
SEROLOGICAL RESEARCH INSTITUTE Xmas Tree Stain
Supplier:  SEROLOGICAL RESEARCH INSTITUTE R540
Encompass
Two dropper bottles of red and green stains. A differential stain for sexual assault evidence slides. Kernechtrot - Picroindigocarmine for finding and differentiating sperm heads and tails. Stains sperm heads red, mid-pieces red and green, tails green and nuclei purple. Store at room temperature.
Catalog No. NC1876722