Learn More
TARGETMOL CHEMICALS INC XMD17-109 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T184210MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. XMD17-109 (ERK5-IN-1) is a new selective ERK-5 inhibitor (EC50 4.2 uM HEK293 cells). purity: 99%
Catalog No. 50-258-1079