Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xp-524 | 2344825-52-9 | 98.0% | C30H28N6O3S | 10MG
SDP

Supplier:  Medchemexpress LLC HY14700810MG

Encompass_Preferred

XP-524 is a small-molecule inhibitor of BET family proteins and EP300, reported to exhibit tumoricidal activity in vivo and to prevent KRAS-driven neoplastic transformation in preclinical models. It has been used to enhance antigen presentation and promote recruitment of cytotoxic T cells, and is supplied with solubility and storage guidance for research use.

  • Potent dual BET and EP300 inhibition demonstrated in preclinical studies.
  • Shows tumoricidal activity and prolongs survival in pancreatic cancer models.
  • Enhances antigen presentation and promotes cytotoxic T-cell tumor recruitment.
  • High reported purity suitable for research applications.
  • Soluble in DMSO for in vitro assays with reported formulation protocols for in vivo use.
  • Recommended storage conditions provided to support compound stability.

Catalog No. 50-002-58473


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.