Learn More
Medchemexpress LLC XP-524 | 2344825-52-9 | 98.02% | 552.65 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY147008
Encompass_Preferred
XP-524 is a potent BET and EP300 inhibitor, demonstrating significant tumoricidal activity in vivo. It prevents KRAS-induced neoplastic transformation and extends survival in aggressive PDAC transgenic mouse models. Additionally, it enhances the presentation of self-peptide and the recruitment of cytotoxic T lymphocytes, making it a promising compound for research into pancreatic ductal adenocarcinoma (PDAC).
  • Potent BET and EP300 inhibitor
  • Exhibits tumoricidal activity in vivo
  • Prevents KRAS-induced neoplastic transformation
  • Extends survival in PDAC mouse models
  • Enhances self-peptide presentation
  • Recruits cytotoxic T lymphocytes
Catalog No. 50-003-62775