Learn More
Medchemexpress LLC XP-59 | 890402-73-0 | 99.7% | 282.30 | 100 MG

Supplier: Medchemexpress LLC HY136284100MG
XP-59 is a potent inhibitor of the SARS-CoV Mpro, demonstrating a Ki of 0.1 μM and 0.1 nM against SARS-CoV Mpro. Its chemical formula is C15H14N4O2, with a molecular weight of 282.30 and CAS No. 890402-73-0. The product is provided as a solid, appearing off-white to light yellow, and has a purity of 99.67%. It is intended for research use only.
- Potent inhibitor of SARS-CoV Mpro with a Ki of 0.1 μM
- Exhibits a Ki of 0.1 nM against SARS-CoV Mpro
- Provided as a solid with an off-white to light yellow appearance
- Purity of 99.67%
- Intended for research use only
- Store powder at -20°C for up to 3 years or 4°C for up to 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.