Learn More
Medchemexpress LLC XP-59 | 890402-73-0 | 99.7% | 282.30 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13628450MG
Encompass_Preferred
XP-59 is a potent inhibitor of the SARS-CoV Mpro, with a Ki of 0.1 μM. The SARS-CoV main proteinase (Mpro) plays a central role in the formation of the viral replicase/transcriptase complex, making it an ideal target for drug development. This product is for research use only.
  • Potent inhibitor of SARS-CoV Mpro
  • Ki: 0.1 nM (SARS-CoV Mpro)
  • Solid appearance
  • Off-white to light yellow color
  • Stable under recommended storage conditions
Catalog No. 50-003-52301