Learn More
Medchemexpress LLC XP-59 | 890402-73-0 | 99.7% | C15H14N4O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13628410MG
Encompass_Preferred
A small-molecule inhibitor of the SARS coronavirus main protease (Mpro) reported with a Ki of 0.1 μM. The compound has molecular formula C15H14N4O2, is provided for research use, and is reported at high purity with recommended dissolution in DMSO.
  • Potent Mpro inhibition (Ki ≈ 0.1 μM) for enzymatic assays.
  • High purity (~99.7%) suitable for biochemical studies.
  • Well-defined chemical identity and formula (C15H14N4O2).
  • Soluble in DMSO for stock preparation.
  • Supplied with a datasheet containing characterization and purity data.
Catalog No. 50-002-61574