Learn More
McKesson General Medical Cepheid Xpert Xpress CoV-2 Plus, Rapid Results In 20 Minutes, FDA EUA Approved, 10 Tests Per Kit
Supplier:  McKesson General Medical 1212776
Encompass_Preferred
The Cepheid Xpert Xpress CoV-2 plus is an automated real-time RT-PCR test for rapid SARS-CoV-2 detection, providing results in as little as 20 minutes. It features a multi-gene target approach for enhanced reliability and versatility in upper respiratory sample processing. Its streamlined workflow minimizes hands-on time to less than 1 minute, making it ideal for efficient laboratory and point-of-care use.
Catalog No. NC2680996