Learn More
ABclonal Technology XPNPEP1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15327500UL
Encompass_Preferred
This gene encodes the cytosolic form of a metalloaminopeptidase that catalyzes the cleavage of the N-terminal amino acid adjacent to a proline residue. The gene product may play a role in degradation and maturation of tachykinins, neuropeptides, and peptide hormones. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-05133