Learn More
XpressBio XpressMyco Master PCR Detection Kit, 50 Reactions
SDP
Supplier:  XpressBio XMYC250
Encompass_Preferred
The XpressMyco Master PCR Detection Kit is designed for the routine examination of cell cultures for mycoplasmas. With few pipetting steps for the PCR approach and analysis by means of gel electrophoresis, the procedure delivers results quickly, easily, and to a high level of sensitivity.
Catalog No. NC2956728