Learn More
Medchemexpress LLC XR8-89 | 2817811-16-6 | C29H36N4O2S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY150784
Encompass_Preferred
XR8-89 is a potent papain-like protease (PLpro) inhibitor with an IC50 value of 0.1 μM. It induces conformational changes in SARS-CoV-2 papain-like protease, inhibiting SARS-CoV-2 replication and can be used for SARS-CoV-2 research.
  • Potent papain-like protease (PLpro) inhibitor
  • Induces conformational changes in SARS-CoV-2 papain-like protease
  • Inhibits SARS-CoV-2 replication
  • Suitable for SARS-CoV-2 research
Catalog No. 50-003-74314