Learn More
New England Biolabs, Inc. XRN-1 – 100 units
SDP
Supplier:  New England Biolabs, Inc. M0338L
Encompass
XRN-1 is highly processive 53 exoribonuclease, requiring 5 monophosphate. It also acts on 5' monophosphate ssDNA with greatly reduced efficiency.
Catalog No. 50-591-119