Learn More
Anton Paar Chemically resistant peristaltic pump for DMA 501/1001
Supplier:  Anton Paar 187548
Encompass_Preferred
Peristaltic pump for single determination of fluids up to 3,000 mPas. Resistant to aggressive mediums.Suitable for use with Anton Paar density metersDMA 501 Compact density and concentration meter NC1818227,DMA 1001 Precision compact density and concentration meter NC1818230
Catalog No. NC1818337