Learn More
Waters Corp Waters Column Kit, XSelect Peptide CSH C18, 130 Å, 2.5 μm, 4.6 mm, 150 mm
Supplier: Waters Corp 176003093
This column kit, featuring CSH130 C18 particles, offers excellent peak shape for peptides with FA- or TFA-containing mobile phases. The column’s performance is robust for LC or LC-MS applications due to low dependence on strong ion pairing agents. Packed in ultra-low dispersion hardware, XSelect CSH130 C18 2.5 μm eXtended Performance [XP] columns ensure exceptional separation, robustness, and throughput for HPLC, enabling seamless transition to UPLC. A Cytochrome C Digestion Standard is included for immediate benchmarking and continued monitoring.
- Unmatched selectivity and consistency for chromatographic analysis.
- Superior separation performance, robustness, and throughput for HPLC assays.
- Compatible with all HPLC, UPLC, and UHPLC technologies.