Learn More
Waters Corp WATERS CORPORATION Column, XSelect CSH C18, 2.5 μm, 3 mm X 150 mm
Supplier: Waters Corp 176002892
XSelect CSH C18 columns are general-purpose C18 columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology enables superior peak shape and increased loading capacity for basic compounds.
- Chemistry: C18, reversed phase
- Particle type: Hybrid, spherical, 2.5 μm
- Pore size: 130 Å
- Dimensions: 3 mm inner diameter, 150 mm length
- pH stability: 1 to 11 pH
- Maximum pressure: 18000 psi (1240 Bar)
- Carbon load: 15%
- Provides exceptional separation performance, robustness, and throughput for HPLC assays.
- Delivers high analyte loading and operational stability across UPLC, HPLC, and preparative separation platforms.
- Ensures consistent results and diverse selectivity.
- Improves HPLC productivity through faster analysis and reduced solvent consumption.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.