Learn More
Waters Corp Waters Column, CSH C18, 3.5 μm, 4.6 mm ID, 75 mm Length
Supplier:  Waters Corp 186005268
Encompass_Preferred
CSH C18 columns are general-purpose columns offering alternative selectivity. They incorporate Charged Surface Hybrid (CSH) technology to address issues like peak shape asymmetry, poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Facilitate high-resolution separations for complex samples.
  • Compatible with all HPLC, UPLC, and UHPLC technologies.
  • Provide consistent batch-to-batch performance.
  • Offer superior performance and comparable backpressure to core-shell columns.
  • CSH technology provides improved loading capacity and particle size scalability.
  • Enhance liquid chromatography performance in acidic, low ionic strength mobile phases.
  • Feature ultra-low dispersion hardware for superb separation.
Catalog No. 50-791-306