Learn More
Waters Corp XSelect CSH C18 Method Validation Kit, 130Å, 5 μm, 4.6 mm x 150 mm, 3 pk
Supplier:  Waters Corp 186005547
Encompass_Preferred
This method validation kit provides three high-performance HPLC columns, each meticulously packed with a distinct gel lot of stationary phase material. Leveraging advanced CSH Technology, these columns deliver an expansive range of selectivities and demonstrate superior performance, particularly in acidic, low ionic strength, and LC/MS friendly mobile phase conditions.
  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 11
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 5 μm particle size
  • 130 Å pore size
  • Designed for HPLC systems
  • CSH particle technology
  • USP classification L1
  • 4.6 mm inner diameter
  • 150 mm length
  • 15% carbon load
  • Three units per package
Catalog No. 50-791-504